missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4515, Orange
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts such as asbestos fibre
- Made from highly breathable SMS non-woven for effective release of body heat and moisture and reduced heat stress
- Soft and strong 50 gsm SMS for strong performance
- 2 panel hood, 2-way zip and storm flap with elasticated back and ankles to reduce the risk of skin exposure
- CE Cat III Type 5/6 hazardous dusts and limited liquid splashes
- EN 1073-2 nuclear particulate hold out

Specifications
Specifications
Product Type | Protective Coveralls |
Size | 4X-Large |
Gender | Unisex |
Material | SMS Polypropylene |
Color | Orange |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Hood Type | 2-panel |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Videos
-
3M Safety Solutions - Protective coveralls overview
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Protective Coverall 4515, Orange >
Spot an opportunity for improvement?Share a Content Correction