missing translation for 'onlineSavingsMsg'
Learn More
3M™ Protective Coverall 4515, Orange Product Code.: 14850982
Add to basket

3M™ Protective Coverall 4515, Orange

Product Code. 14850982
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 14850982

Brand: 3M™ 7000089632

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Strong 50 gsm SMS coverall offers breathability and comfort to be worn comfortably for long periods of time. Protects against toxic dusts Type 5, liquid splashes Type 6, and nuclear particles to EN 1073-2. Available in a variety of colours to assist in identification and zoning.

  • Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts such as asbestos fibre
  • Made from highly breathable SMS non-woven for effective release of body heat and moisture and reduced heat stress
  • Soft and strong 50 gsm SMS for strong performance
  • 2 panel hood, 2-way zip and storm flap with elasticated back and ankles to reduce the risk of skin exposure
  • CE Cat III Type 5/6 hazardous dusts and limited liquid splashes
  • EN 1073-2 nuclear particulate hold out
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Small
Gender Unisex
Material SMS Polypropylene
Color Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class PPE Directive Category III, Type 5/6
Size (English) Chest 33 to 36 in.
Size (Metric) Chest 84 to 92 cm
Height (English) 64 to 67 in.
Height (Metric) 164 to 170 cm
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      3M Safety Solutions - Protective coveralls overview

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    3M™ Protective Coverall 4515, Orange >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.