missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Orange Disposable Protective Coverall 4515
Protective Coverall 4515 Series protect from hazardous dusts such as asbestos fibers.
£159.00
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | SMS Polypropylene |
Color | Orange |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11993764
|
3M™
7000089638 |
4X-Large |
£159.00
Pack of 20 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
11993744
|
3M™
7000089633 |
Medium |
£159.00
Pack of 20 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
14870982
|
3M™
7000089634 |
Large |
£159.00
Pack of 20 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11913764
|
3M™
7000089636 |
2X-Large |
£159.00
Pack of 20 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
14810992
|
3M™
7000089637 |
3X-Large |
£159.00
Pack of 20 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
11973754
|
3M™
7000089635 |
X-Large |
£159.00
Pack of 20 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
14850982
|
3M™
7000089632 |
Small |
£159.00
Pack of 20 |
In Stock
Log in to see stock. |
|||||
Description
- Breathable material reduces heat build-up and discomfort
- Excellent barrier against dry particles and limited liquid splash (CE Type 5/6)
- Robust, highly breathable 50 gsm SMS material
- Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
- Two-way zipper with fully sealable storm flap
- Two-panel hood design
- Elasticated wrists, waist, and ankles for increased comfort and freedom of movement
Specifications
General Purpose Coveralls | |
SMS Polypropylene | |
Single-use | |
Elastic | |
2-panel | |
Serged | |
PPE Directive Category III, Type 5/6 |
Unisex | |
Orange | |
Elastic | |
Elastic | |
Front Zipper with Storm Flap | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title