Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Product Code. 11736003
Click to view available options
Filter Efficiency:
FFP1 NR D
FFP2
FFP2 NR D
With Exhalation Valve:
No
Yes
Quantity:
1 Ea.
10/Pk.
20/Pk.
Unit Size:
Pack of 10
Pack of 20
Product Code. 11736003
Brand: 3M™ 7100010817
Only null left
Add to basket
Add to basket
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance European Community Directive 89/686/EEC, EN 149:2001 A1:2009, CE
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP2 NR D
Product Type General Purpose Particulate Respirator
With Exhalation Valve Yes
Filter Efficiency FFP2 NR D
For Use With (Application) Protection against dust and mist
Latex Free Yes
Model 8822
Quantity 10/Pk.
Resistant To Dusts, mists and Metal fumes
Shape Convex
Sterility Non-sterile
FDA Cleared Surgical Mask No