missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
Brand: 3M™ 7000034734
13.50 GBP valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.
Specifications
European Community Directive 89/686/EEC, EN 149:2001 A1:2009, CE | |
Single-use | |
General Purpose Particulate Respirator | |
FFP1 NR D | |
Yes | |
20/Pk. | |
Convex | |
No |
Cup/Molded | |
FFP1 NR D | |
No | |
Protection against dust and mist | |
8710 | |
Dusts, mists and Metal fumes | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Disposable Particulate Respirator, 8000 Series > Filter Type: FFP1 NR D; With valve; Qty: 20 Pack
Spot an opportunity for improvement?Share a Content Correction