missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable PES Bottle Top Filters

Increase sample protection with Fisherbrand Sterile Disposable Bottle Top Filters with PES Membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

Brand:  Fisherbrand™

72.97 GBP valid until 2024-07-31
Use promo code "21836" to get your promotional price.



Additional Details : Weight : 1.44000kg

 View more versions of this product

Product Code. 15983307

  • £139.32 / Pack of 12
In Stock
to see stock.
Add to basket

Alert:

CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.


Description

Description

  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Marked with catalog number, membrane type, pore size, lot number and expiration date for easy traceability
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • Bottom screw threads for attachment to bottles with 45 mm neck sizes
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal

Certifications

Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility

Specifications

Specifications

Non-pyrogenic per USP Standard Lot-to-Lot
PES
Sterile
Bottle Top Unit
75 mm
Single-use
0.45 μm
Sterile Filtration of media and serum
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.