Learn More
Fisherbrand™ Disposable PES Filter Units
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
Brand: Fisherbrand™
148.13 GBP valid until 2024-07-31
Use promo code "21836" to get your promotional price.
Additional Details : Weight : 4.25700kg
Includes: Individually packaged sterile screw top style bottle cap
Description
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
- PES membrane is low protein binding to maintain protein balance
- PES membrane has low extractables to maintain media purity
- Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date
- Non-pyrogenic and non-cytotoxic
- Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
- 0.2μm units for sterile filtration
- 0.45μm units for fluid clarification and particle removal
Certifications
Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility
Specifications
Low binding | |
Single-use | |
0.45 μm | |
Fluid clarification and particle removal | |
90 mm |
Non-pyrogenic per USP Standard Lot-to-Lot | |
PES | |
Sterile | |
1000 mL |