Learn More
Nexcelom Bioscience Disposable Plastic Cell Counting Chambers
Disposable, standard slide thickness cell counting chamber slide. Nexcelom™ Disposable Plastic Cell Counting Chambers are designed with two counting chambers per slide.
Brand: Nexcelom Bioscience CHT4-SD100-002
Additional Details : Weight : 0.16920kg
Description
Disposable, standard slide thickness cell counting chamber slide. Nexcelom™ Disposable Plastic Cell Counting Chambers are designed with two counting chambers per slide.
- Standard slide thickness
- For cells ranging from 2 to 80μm in diameter
- Each slide has a protective film on both sides that must be removed before use
- Cost affective
- Disposable
- Convenient
- Two counting chambers per slide
- Box of 75 slides for 150 counts
Specifications
Cell Counting Chamber | |
Plastic | |
75 |