missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Disposable Polypropylene Griffin Low-form Beakers

Molded-in stacking ribs prevent beakers from sticking together when stacked

Brand:  Cole-Parmer™ 06006-83

Product Code. 12323840

  • £60.05 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Disposable polypropylene Griffin low-form beakers, 600 mL
  • Graduated in mL and oz.
  • Available in packs of 25
  • Autoclavable
Specifications

Specifications

Griffin Low-form Beaker
Flat
600 mL
Clear
4.5 in.
5 in.
Polypropylene
20 oz.
Single-use
25 Beakers
11.4 cm
12.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.