Learn More
New Source™ Disposable Chemical Resistant SMS Coveralls Type 5/6
Product Code. 16655042
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
X-Large
Disposable chemical resistant coveralls offering protection against solid particulates and liquid splashes and sprays. CE certified as PPE Cat III under Regulation EU 2016/425
  • Conforms to CE Category III Type 5/6
  • Non-woven SMS disposable fabric
  • Protects against chemical and biological hazards
Specifications
Size X-Large
Gender Unisex
Material Non-woven SMS
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Flap
Seam Type Stitched
Resistance Particle
Certifications/Compliance CE Cat III PPE meeting: EN ISO 13982-1:2004, EN ISO 13034: 2005 +A1:2009, EN ISO 14325:2004
Description Disposable coverall with elasticated hood, cuffs, waist and ankles. Without shoe cover.
Weight (Metric) 55 gsm