missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Source™ Disposable SPP Shoe Cover with Anti-Slip Sole
Disposable spunbond polypropylene shoe cover, with anti-skid sole. Wear over your shoes in non-hazardous environments to protect against dirt and grime. Ideal for non-critical areas or pre-gowning entry rooms.
Brand: New Source™ HLSCSPP
This item is not returnable.
View return policy
Description
- SPP fabric
- Long stretch: 44 cm (A)
- Heel height: 16 cm (B)
- Weight: 38 gsm
- Blue/White
- PPE Category I
- Non-sterile
Specifications
Universal | |
Blue, White | |
Non-sterile | |
PPE Category I | |
Length: 44 cm; Heel Height: 16 cm |
SPP Fabric | |
Single-use | |
Anti-slip | |
100 Shoe Covers/Bag, 3 Bags/Carton | |
38 gsm |