missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Dissolved Oxygen Water Testing Kit

Measure dissolved oxygen by modified Winkler titration. Hanna Instruments™ Dissolved Oxygen Water Testing Kit is supplied complete with all reagents and equipment necessary to perform approximately 110 tests.

Brand:  Hanna Instruments™ HI3810

Code : 04

Additional Details : Weight : 0.26000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


Product Code. 10470684

  • £271.00 / Each
Estimated Shipment: 19-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Manual titration performed with starch indicator
Specifications

Specifications

110
0 to 10mg/L
Dissolved Oxygen Kit
1 Each
Modified Winkler
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dissolved oxygen test kit (110 tests average)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P390-Absorb spillage to prevent material damage.
  • P391-Collect spillage.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, potassium iodide, sodium azide, manganese(II) sulphate monohydrate, sulphuric acid

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers