missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tristel™ Distel™ Surface Disinfectant
Click to view available options
Volume (Metric):
500 mL
1000 mL
5000 mL
Composition:
Distel TM301
Distel TM305
Distel TM309
Unit Size:
5000mL
500mL
Pack of 1000
Container Type:
Spray bottle
Description
- Classified as a biocide under the EU Biocides Regulation
- Tested and approved as bactericidal, fungicidal, virucidal and tuberculocidal
- Effective in the presence of blood borne diseases (HIV/Hepatitis B) and also destroys DNA and RNA
- Recommended for use at a dilution of 1:100 for all normal laboratory requirements
- Suitable for use on most surfaces including glass, metal, plastic, rubber and fabric
Specifications
Specifications
| Product Type | Surface Disinfectant |
| Form | Concentrate |
| Composition | Distel TM309 |
| Dilution | 0.11111111111 |
| For Use With (Application) | Laboratory equipment, Benches and shelves, Fridges, pipettors, Water baths, Incubators, Centrifuges, Discard jars, Non-critical devices |
| Volume (Metric) | 5000 mL |
Safety and Handling
Product Identifier
- DISTEL HIGH LEVEL LABORATORY DISINFECTANT
- Danger
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction