Learn More
Distillation Standard ASTM D 86; NF EN ISO 3405 (Nominal value: 30-178.6 °C), Paragon Scientific™
Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405
Brand: Paragon Scientific CRM-DIGA
Additional Details : Weight : 0.25000kg Transport : UN number : 1203 Chem class : 3 Pack group : II
Description
- Certified Reference Material for Distillation
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified via Round Robin
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
UN 1203 |
Safety and Handling
- Distillation Standard
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Flammable liquid Category 1
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : gasoline
Nominal values quoted. Certified Values will vary from batch to batch.