missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Distilling Head

Distilling Head made of durable DURAN™ glass tubing.

Brand:  Lenz Laborglasinstrumente™ 05030022

 View more versions of this product

Product Code. 10171471

  • £56.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Meets DIN 12594 standards
  • NS socket and two NS cones
  • Descending angle of 75°
Specifications

Specifications

For distillation thermometers of 55 mm built-In length
Duran Glass
NS 19/26 Vertical, NS 19/26 Oblique
Distilling Head
DIN 12594
Claisen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers