Learn More
Lenz Laborglasinstrumente™ Distilling Links
Distilling Links for the connection of distillation thermometers of 55 mm built-in length.
£39.00 - £161.00
Product Code | Brand | Length (Metric) Jacket | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Jacket | Standard Taper Joint | Price | Quantity & Availability | ||||
10686755
|
Lenz Laborglasinstrumente™
05050211 |
160 mm | NS 14/23 |
£109.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10702682
|
Lenz Laborglasinstrumente™
05053201 |
160 mm | NS 14/23, NS 14/23 |
£63.98
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10147044
|
Lenz Laborglasinstrumente™
05053314 |
250 mm | NS 29/32, GL 14 |
£147.00
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10491865
|
Lenz Laborglasinstrumente™
05050314 |
250 mm | NS 29/32 |
£121.78
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10548081
|
Lenz Laborglasinstrumente™
05053304 |
250 mm | NS 14/23, NS 29/32 |
£71.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10634271
|
Lenz Laborglasinstrumente™
05050304 |
250 mm | NS 29/32, NS 14/23 |
£63.00
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10234341
|
Lenz Laborglasinstrumente™
05040003 |
300 mm | NS 24/29 |
£62.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10363181
|
Lenz Laborglasinstrumente™
05040004 |
350 mm | NS 29/32, NS 29/32 |
£63.65
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10376021
|
Lenz Laborglasinstrumente™
05047004 |
350 mm | NS 14/23, NS 29/32 |
£39.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11816214
|
Lenz Laborglasinstrumente™
05043004 |
350 mm | NS 29/32 |
£133.56
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10545670
|
Lenz Laborglasinstrumente™
05053414 |
400 mm | NS 29/32 |
£161.00
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- With Liebig condenser
- Outlet with NS cone
- Water connection with glass olives, with NS socket
Specifications
Distilling Link | |
Duran Glass |