Learn More
Lenz Laborglasintrumente Distilling Links acc. to Claisen, with GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05055414
Additional Details : Weight : 0.01000kg
Description
- Distilling links with Liebig condenser (straight outlet) and head to Claisen
- Water connection with DIN thread GL14
- With two sockets NS 14/23 for distillation thermometers of 55 mm built-in length
- Made of DURAN™ tubing
Specifications
400 mm | |
NS 29/32 |