Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05305004
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10000491
  • £694.00 / Each
Estimated Shipment: 28-08-2024
to see stock.
Description
Description
  • Receivers with straight adapter
  • With cooling jacket, graduated, with socket and cone NS 29/32
  • Vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass
50 mL
Socket and Cone NS 29/32