missing translation for 'onlineSavingsMsg'
Learn More

Dithiooxamide, 98%

Product Code. 15389252 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15389252

Brand: Thermo Scientific Acros 117550050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-40-3
Molecular Formula C2H4N2S2
Molecular Weight (g/mol) 120.19
InChI Key OAEGRYMCJYIXQT-UHFFFAOYSA-N
Synonym dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
PubChem CID 2777982
IUPAC Name ethanedithioamide
SMILES C(=S)(C(=S)N)N

Specifications

Melting Point 200.0°C
Elemental Analysis 22.7 to 23.9% (N) 19.5 to 20.5% (C)
Infrared Spectrum Authentic
Linear Formula H2NCSCSNH2
Quantity 5 g
Merck Index 15,8412
Solubility Information Solubility in water: 0.37g/L (20°C).
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material Dithiooxamide
compliance-icons
Product Identifier
  • Dithiooxamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.