missing translation for 'onlineSavingsMsg'
Learn More
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals Product Code.: 15875458

Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15875458 Shop All Thermo Scientific Products
5 g, Plastic bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15875458

Brand: Thermo Scientific J15397.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as protective agent for SH groups

Used as a protective agent for SH groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dttShow MoreShow Less
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2 S,3 S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Melting Point 40°C to 43°C
Boiling Point 125°C to 130°C/12 mm
Flash Point >110°C
Packaging Plastic bottle
Quantity 5 g
Beilstein 1719757
Formula Weight 154.25
Percent Purity ≥99.5%
Grade Molecular Biology, Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material Dithiothreitol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number240753 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ15397.06,J15397.22,J15397.03,J15397.14
Lot Number235799 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ15397.06,J15397.22,J15397.03,J15397.14
Lot Number203283 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ15397.06,J15397.22,J15397.03,J15397.14
Lot Number205246 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ15397.06,J15397.22,J15397.03,J15397.14
Lot Number182408 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberJ15397.06,J15397.22,J15397.03,J15397.14
compliance-icons
Product Identifier
  • Dithiothreitol, Molecular Biology Grade, Ultrapure, Thermo Scientific
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-dimercapto-2,3-Butanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.