missing translation for 'onlineSavingsMsg'
Learn More
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™ Product Code.: 10592945

Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™

Product Code. 10592945
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10592945

Brand: Fisher BioReagents BP1725

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

  • Commonly used as reducing agent to disrupt disulfide bonds in proteins in SDS-PAGE
  • Functionally tested for use in electrophoresis
  • Tested for absence of DNase, RNase, and protease
  • DTT is also used as an enzyme stabilizer during protein purification and enzymatic reactions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Absorbance 0.04 AU max.
Melting Point 38°C
Color White
pH 5.1
Boiling Point 125°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 5 g
Merck Index 15, 3419
Formula Weight 154.24
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Physical Form Powder or Solid
Chemical Name or Material Dithiothreitol
Show More Show Less
compliance-icons
Product Identifier
  • DL-1,4-Dithiothreitol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dithiothreitol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.