missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithranol, Honeywell Fluka™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 1143-38-0 |
Molecular Formula | C14H10O3 |
MDL Number | MFCD00053409 |
Quantity | 1 g |
UN Number | NONH for all modes of transport |
Beilstein | 2054360 |
Synonym | anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy |
InChI Key | NUZWLKWWNNJHPT-UHFFFAOYSA-N |
SMILES | OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 |
IUPAC Name | 1,8-dihydroxy-9,10-dihydroanthracen-9-one |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'