missing translation for 'onlineSavingsMsg'
Learn More

Dizocilpine maleate, 99+%, Thermo Scientific Chemicals

Product Code. 15416079
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NMDA receptor antagonist that prevents calcium flux

Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.

Solubility
Soluble in ethanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77086-22-7
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD00082465
InChI Key QLTXKCWMEZIHBJ-PJGJYSAQSA-N
Synonym dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126
PubChem CID 6420042
SMILES CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O

Specifications

Color White
Quantity 50 mg
Merck Index 14,3386
Solubility Information Soluble in ethanol.
Formula Weight 337.37
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Dizocilpine maleate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.