Learn More
Velp Scientifica™ DK 20 Digestion Unit
Kjeldahl Digestion Unit with a 20-position aluminum heating block that offers an excellent thermal homogeneity.
Brand: Velp Scientifica™ S30100440
Description
- The DK 20 series perform batch digestion of up to 20 samples with 250 mL test tubes - 42 mm dia.
- The DK 20 is a versatile digester suitable for Kjeldahl N/Protein applications and Chemical Oxygen Demand in the environmental industry.
- To use the DK 20 for COD (Chemical Oxygen Demand) is necessary to configure it by adding a different support system able to manage all the glass required for the COD analysis for processing 20 samples simultaneously.
- For applications where the digestion process produces fumes, acid gases or corrosive substances, the KS 1000 Scrubber is strongly recommended.
EXCELLENT ACCURACY AND REPEATABILITY:
The aluminium heating block ensures high thermal homogeneity heating up to 450°C.
- Temperature selection with 1°C steps and a precision of ± 0.2°C.
- Heating block temperature stability of ± 0.5°C.
ENERGY SAVINGS:
The DK Series digesters guarantee fast temperature set point reaching, reducing energy consumption.
- Average 13% lower consumption compared to the previous models.
SPACE SAVING DESIGN:
Save valuable space under the fume hood with the VELP DK digesters. The design of the DK Series ensures minimum space requirements.
INTUITIVE INTERFACE:
The bright digital display provides maximum visibility and easy reading of the information.
- Supports the recording of up to 20 methods with 4 temperature ramps.
- Easy to use interface with 4 buttons.
- The inclination of the front panel protects it from chemical spillage at high temperatures.
VERSATILITY:
DK models are available with tubes and all necessary parts to operate.
- A wide range of accessories is available to match analytical requirements.
- Genuine consumables to digest the samples efficiently.
Specifications
Digital | |
230 V | |
20 x 250 mL | |
Instrument and Mandatory Accessories | |
20 | |
450°C |
50/60 Hz | |
20 x 8.5 oz. | |
5100 x 330 x 140 mm | |
Stainless Steel, Epoxy Painted with Chemical Resistant Coating | |
842°F |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.