missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

Brand:  Thermo Scientific Chemicals 426382500

Code : 29

Additional Details : CAS Number : 3483-12-3 Weight : 0.26000kg


 View more versions of this product

Product Code. 10215054

  • £345.00 / 250mL
In Stock
to see stock.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Biochemistry
Specifications

Specifications

DL-1, 4-Dithiothreitol
1M Solution in water, For molecular biology
C4H10O2S2
0.90 to 1.10M
15, 3419
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.24
Glass bottle
250 mL
Colorless
3483-12-3,7732-18-5
MFCD00004877
1.0400g/mL
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
C(C(C(CS)O)O)S
154.24
CHEBI:42170
Biochemical
1.04
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DL-Dithiothreitol, 1M solution in water

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only