missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Brand: Thermo Scientific Chemicals 426380500
71.95 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For BiochemistrySpecifications
DL-1, 4-Dithiothreitol | |
1M Solution in water, For molecular biology | |
C4H10O2S2 | |
0.90 to 1.10M | |
15, 3419 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol | |
446094 | |
154.24 | |
Glass bottle | |
50 mL |
Colorless | |
3483-12-3, 7732-18-5 | |
MFCD00004877 | |
1.0400g/mL | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
C(C(C(CS)O)O)S | |
154.24 | |
CHEBI:42170 | |
Biochemical | |
1.04 | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- DL-Dithiothreitol, 1M solution in water
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only