missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Isopropylserine, 99%, Thermo Scientific™
£24.80 - £74.70
Chemical Identifiers
CAS | 7522-43-2 |
---|---|
Molecular Formula | C6H13NO3 |
Molecular Weight (g/mol) | 147.17 |
MDL Number | MFCD01863303 |
InChI Key | QTYSAGIKKRDGIV-ZCFIWIBFSA-N |
Synonym | dl-2-isopropylserine, s-2-amino-2-hydroxymethyl-3-methylbutanoic acid, 2-isopropyl-l-serine, 2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid |
PubChem CID | 7010084 |
IUPAC Name | (2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid |
SMILES | CC(C)C(CO)(C(=O)O)N |
Description
Chemical Identifiers
7522-43-2 | |
147.17 | |
QTYSAGIKKRDGIV-ZCFIWIBFSA-N | |
7010084 | |
CC(C)C(CO)(C(=O)O)N |
C6H13NO3 | |
MFCD01863303 | |
dl-2-isopropylserine, s-2-amino-2-hydroxymethyl-3-methylbutanoic acid, 2-isopropyl-l-serine, 2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid | |
(2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid |
Specifications
7522-43-2 | |
Cream | |
99% | |
MFCD01863303 | |
Solubility in water: .. Other solubilities: soluble in aq. acids and bases | |
CC(C)C(CO)(C(=O)O)N | |
147.17 | |
147.17 | |
Crystalline Powder |
262°C | |
Authentic | |
C6H13NO3 | |
dl-2-isopropylserine, s-2-amino-2-hydroxymethyl-3-methylbutanoic acid, 2-isopropyl-l-serine, 2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid | |
QTYSAGIKKRDGIV-ZCFIWIBFSA-N | |
(2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid | |
7010084 | |
99% | |
DL-2-Isopropylserine, 99% |
RUO – Research Use Only