missing translation for 'onlineSavingsMsg'
Learn More
DL-2-Pentanol, 98%, Thermo Scientific Chemicals Product Code.: 10397150

DL-2-Pentanol, 98%, Thermo Scientific Chemicals

Product Code. 10397150
1 L, Glass bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10397150

Brand: Thermo Scientific Acros 129980010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6032-29-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004579
InChI Key JYVLIDXNZAXMDK-UHFFFAOYSA-N
Synonym 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
PubChem CID 22386
ChEBI CHEBI:77518
IUPAC Name pentan-2-ol
SMILES CCCC(C)O

Specifications

Melting Point -50°C
Color Colorless
Density 0.8120g/mL
Boiling Point 118°C to 119°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(OH)CH3
Refractive Index 1.4050 to 1.407
Quantity 1 L
Beilstein 01, 384
Residue after Evaporation 0.003% max.
Merck Index 15, 7231
Specific Gravity 0.812
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with alcohol and ether
Viscosity 4.1 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Pentanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464176 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number129980010,129985000,129980025
Lot NumberA0456722 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number129980010,129985000,129980025
Lot NumberA0452576 Certificate TypeCertificate of Analysis Date27/04/2023 Catalog Number129980010,129985000,129980025
Lot NumberA0394093 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number129980010,129985000,129980025
Lot NumberA0404263 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number129980010,129985000,129980025
compliance-icons compliance-icons
Product Identifier
  • DL-2-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-2-Pentanol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.