missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Arginine, 98%
Click to view available options
Quantity:
5 g
25 g
Description
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼230°C (decomposition) |
Beilstein | 1725411 |
Solubility Information | Insoluble in water. |
Formula Weight | 174.2 |
Percent Purity | 98% |
Quantity | 5 g |
Sensitivity | Air Sensitive |
Chemical Name or Material | DL-Arginine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'