missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-beta-Homoproline hydrochloride, 97%

Product Code. 15484367
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15484367

Brand: Thermo Scientific Chemicals H31749.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71985-79-0 | C6H12ClNO2 | 165.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71985-79-0
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD11506301
InChI Key VQDACVOAOJQTPR-UHFFFAOYSA-N
Synonym pyrrolidin-2-yl-acetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hydrochloride, pyrrolidin-2-ylacetic acid hydrochloride, 2-pyrrolidineacetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hcl, h-d-beta-hopro-oh.hcl, 2-pyrrolidineacetic acid hcl, dl-beta-homoproline hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2-pyrrolidinylacetic acid hydrochloride
PubChem CID 12514707
IUPAC Name 2-pyrrolidin-2-ylacetic acid;hydrochloride
SMILES C1CC(NC1)CC(=O)O.Cl

Specifications

Melting Point 171°C to 174°C
Quantity 250 mg
Formula Weight 165.62
Percent Purity 97%
Chemical Name or Material DL-beta-Homoproline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.