missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Camphor, Extra Pure, SLR, Synthetic, Fisher Chemical
Click to view available options
250g, Glass Jar
Specifications
Specifications
CAS | 76-22-2 |
Melting Point | 175°C |
Color | White |
Boiling Point | 204°C |
Packaging | Glass Jar |
Molecular Formula | C10H16O |
MDL Number | 74738 |
Quantity | 250 g |
InChI Key | DSSYKIVIOFKYAU-UHFFFAOYNA-N |
SMILES | CC1(C)C2CCC1(C)C(=O)C2 |
Show More |
Safety and Handling
Product Identifier
- DL-Camphor
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction