missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine, 96%, Thermo Scientific Chemicals

Catalog Number p-7037402
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3374-22-9
C3H7NO2S
121.15
MFCD00004881
XUJNEKJLAYXESH-UHFFFAOYNA-N
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
594
CHEBI:15356
2-amino-3-sulfanylpropanoic acid
NC(CS)C(O)=O
This item is not returnable. View return policy
3374-22-9
C3H7NO2S
121.15
MFCD00004881
XUJNEKJLAYXESH-UHFFFAOYNA-N
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
594
CHEBI:15356
2-amino-3-sulfanylpropanoic acid
NC(CS)C(O)=O

DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 225°C (decomposition)
Beilstein 1721406
Solubility Information Soluble in water (partly),1 N HCl (25mg/ml),ethanol,and acetic acid. Insoluble in acetone.
Formula Weight 121.16
Percent Purity 96%
Quantity 5 g
Chemical Name or Material DL-Cysteine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'