missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Dithiothreitol, 99.5%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Specifications
Specifications
Color | White |
Assay Percent Range | >99% |
Quantity | 10 g |
Formula Weight | 154.2 |
Grade | Molecular Biology Reagent |
Physical Form | Powder |
Chemical Name or Material | DL-Dithiothreitol |
Safety and Handling
Product Identifier
- DL-Dithiothreitol, molecular biology reagent
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : DL-Dithiothreitol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction