missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Dithiothreitol, Tocris Bioscience™
Click to view available options
Quantity:
10 g
Description
Reagent for maintaining SH groups in the reduced state.
Specifications
CAS | 3483-12-3 |
Molecular Formula | C4H10O2S2 |
Quantity | 10 g |
Synonym | dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt |
InChI Key | VHJLVAABSRFDPM-IMJSIDKUSA-N |
SMILES | C(C(C(CS)O)O)S |
IUPAC Name | (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
Molecular Weight (g/mol) | 154.24 |
PubChem CID | 446094 |
ChEBI | CHEBI:42170 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'