missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11483308
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19285-83-7
Molecular Formula C5H11NO5
Molecular Weight (g/mol) 165.145
MDL Number MFCD00150703
InChI Key OZDAOHVKBFBBMZ-UHFFFAOYSA-N
Synonym dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
PubChem CID 16219385
IUPAC Name 2-aminopentanedioic acid;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 4155097
Merck Index 14,4469
Density 1.46
Quantity 100 g
Formula Weight 165.15 (147.13 Anhydrous)
Percent Purity 99%
Chemical Name or Material DL-Glutamic acid monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'