missing translation for 'onlineSavingsMsg'
Learn More
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals Product Code.: 15284487

DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals

Product Code. 15284487
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15284487

Brand: Thermo Scientific Acros 450710010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23147-59-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key HQAMJDIBKAKYMU-UHFFFAOYSA-N
Synonym 3,6-bis hydroxymethyl-1,4-dioxane-2,5-diol, pubchem10426, dl-glyceraldehyde dimer+, 2,5-dihydroxy-3,6-dihydroxymethyl-1,4-dioxane
PubChem CID 251615
IUPAC Name 3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
SMILES C(C1C(OC(C(O1)O)CO)O)O

Specifications

Melting Point 138.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.16
Percent Purity 95%
Chemical Name or Material DL-Glyceraldehyde dimer
Product Identifier
  • DL-Glyceraldehyde dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.