missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Homocysteine thiolactone hydrochloride, 99%, Thermo Scientific Chemicals
£23.50 - £53.90
Chemical Identifiers
CAS | 6038-19-3 |
---|---|
Molecular Formula | C4H8ClNOS |
Molecular Weight (g/mol) | 153.624 |
MDL Number | MFCD00012724 |
InChI Key | ZSEGSUBKDDEALH-UHFFFAOYSA-N |
Synonym | dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride |
PubChem CID | 110753 |
IUPAC Name | 3-aminothiolan-2-one;hydrochloride |
SMILES | C1CSC(=O)C1N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357216
|
Thermo Scientific Alfa Aesar
L09077.14 |
25 g |
£23.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367216
|
Thermo Scientific Alfa Aesar
L09077.22 |
100 g |
£53.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.
Solubility
Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
6038-19-3 | |
153.624 | |
ZSEGSUBKDDEALH-UHFFFAOYSA-N | |
110753 | |
C1CSC(=O)C1N.Cl |
C4H8ClNOS | |
MFCD00012724 | |
dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride | |
3-aminothiolan-2-one;hydrochloride |
Specifications
6038-19-3 | |
C4H8ClNOS | |
3562103 | |
dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride | |
ZSEGSUBKDDEALH-UHFFFAOYSA-N | |
3-aminothiolan-2-one;hydrochloride | |
110753 | |
99% |
∽204°C (decomposition) | |
MFCD00012724 | |
14,4734 | |
Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol. | |
C1CSC(=O)C1N.Cl | |
153.624 | |
153.63 | |
DL-Homocysteine thiolactone hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 227-923-3
missing translation for 'rtecsNumber' : XN1928400
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only