missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Lactic acid, 85+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Assay Percent Range | 85+% |
Linear Formula | CH3CH(OH)CO2H |
Merck Index | 15, 5384 |
Formula Weight | 90.08 |
Percent Purity | 85.0 to 90.0% |
Grade | ACS Reagent |
Density | 1.2000g/mL |
Quantity | 500 g |
Packaging | EcoSafPak™ Glass Bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'