missing translation for 'onlineSavingsMsg'
Learn More
DL-Malic acid, 98%, Thermo Scientific Chemicals Product Code.: 11428960

DL-Malic acid, 98%, Thermo Scientific Chemicals

Product Code. 11428960
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 11428960

Brand: Thermo Scientific Alfa Aesar A17874.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

Solubility
Soluble in acetone, ether, water, methanol and ethanol.

Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6915-15-7
Melting Point 130°C to 133°C
Assay Percent Range 0.98
Molecular Formula C4H6O5
MDL Number MFCD00064212
Beilstein 1723539
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid
Solubility Information Soluble in acetone,ether,water,methanol and ethanol.
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILES OC(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxybutanedioic acid
Molecular Weight (g/mol) 134.09
PubChem CID 525
ChEBI CHEBI:6650
Formula Weight 134.09
Density 1.609
Quantity 250 g
Flash Point 203°C
Chemical Name or Material DL-Malic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10099191 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberA17874.30,A17874.0E,A17874.36
Lot Number10249889 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA17874.30,A17874.0E,A17874.36
Lot Number10247979 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberA17874.30,A17874.0E,A17874.36
Lot Number10227555 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog NumberA17874.30,A17874,A17874.0E,A17874.36
Lot Number10236187 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberA17874.30,A17874,A17874.0E,A17874.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Malic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.