missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 117°C to 121°C |
Color | White |
Assay Percent Range | 99+% |
Linear Formula | C6H5CH(OH)CO2H |
Formula Weight | 152.15 |
Percent Purity | ≥99.0% (Acid) |
Quantity | 100 g |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Physical Form | Crystals or Crystalline Powder |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0398912 | Certificate of Analysis | 14/12/2022 | 12531018,125310050,125312500,125310010,125311000 |
A0393328 | Certificate of Analysis | 14/12/2022 | 12531018,125310050,125312500,125310010,125311000 |
A0404971 | Certificate of Analysis | 13/12/2022 | 12531018,125310050,125312500,125310010,125311000 |
A0416997 | Certificate of Analysis | 12/12/2022 | 12531018,125310050,125312500,125310010,125311000 |
A0407657 | Certificate of Analysis | 29/08/2022 | 12531018,125310050,125312500,125310010,125311000 |
Safety and Handling

Product Identifier
- DL-Mandelic acid
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction