missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%
Click to view available options
Quantity:
5 g
25 g
Description
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
Solubility
Soluble in water and Hcl.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
Solubility
Soluble in water and Hcl.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼230°C (decomposition) |
Solubility Information | Soluble in water and Hcl. |
IUPAC Name | 2-amino-4-methylsulfinylbutanoic acid |
Formula Weight | 165.21 |
Percent Purity | ≥98% |
Quantity | 5 g |
Chemical Name or Material | DL-Methionine sulfoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'