missing translation for 'onlineSavingsMsg'
Learn More

DL-Norleucine, 98%, Thermo Scientific Chemicals

Product Code. 11494290
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-06-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064422
InChI Key LRQKBLKVPFOOQJ-UHFFFAOYNA-N
Synonym dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid
PubChem CID 9475
ChEBI CHEBI:36405
IUPAC Name 2-aminohexanoic acid
SMILES CCCCC(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721748
Merck Index 14,6706
Formula Weight 131.18
Percent Purity 98%
Quantity 500 g
Odor Odorless
Chemical Name or Material DL-Norleucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'