missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Norleucine, 98%, Thermo Scientific Chemicals
£46.40 - £579.00
Chemical Identifiers
CAS | 616-06-8 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00064422 |
InChI Key | LRQKBLKVPFOOQJ-UHFFFAOYNA-N |
Synonym | dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid |
PubChem CID | 9475 |
ChEBI | CHEBI:36405 |
IUPAC Name | 2-aminohexanoic acid |
SMILES | CCCCC(N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474290
|
Thermo Scientific Alfa Aesar
A10791.14 |
25 g |
£46.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484290
|
Thermo Scientific Alfa Aesar
A10791.22 |
100 g |
£155.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494290
|
Thermo Scientific Alfa Aesar
A10791.36 |
500 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
616-06-8 | |
131.18 | |
LRQKBLKVPFOOQJ-UHFFFAOYNA-N | |
9475 | |
2-aminohexanoic acid |
C6H13NO2 | |
MFCD00064422 | |
dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid | |
CHEBI:36405 | |
CCCCC(N)C(O)=O |
Specifications
>300°C | |
C6H13NO2 | |
1721748 | |
dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid | |
CCCCC(N)C(O)=O | |
131.18 | |
CHEBI:36405 | |
98% | |
DL-Norleucine |
616-06-8 | |
MFCD00064422 | |
14,6706 | |
LRQKBLKVPFOOQJ-UHFFFAOYNA-N | |
2-aminohexanoic acid | |
9475 | |
131.18 | |
Odorless |
Safety and Handling
missing translation for 'einecsNumber' : 210-462-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only