missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Phenylalanine, 99% Product Code.: 11499382

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Product Code. 11499382
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11499382

Brand: Thermo Scientific Chemicals A10132.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∼266°C (decomposition)
Beilstein 1910407
Merck Index 14,7271
Solubility Information Soluble in water. Insoluble in ethyl ether,benzene.
Quantity 25 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material DL-Phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245672 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberA10132.22,A10132.14,A10132.36
Lot Number10241697 Certificate TypeCertificate of Analysis Date05/07/2022 Catalog NumberA10132.22,A10132.14,A10132.36
Lot NumberH6889A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10132.22,A10132.14,A10132.36
Lot NumberJ6655A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10132.22,A10132.14,A10132.36
Lot NumberJ1690A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10132.22,A10132.14,A10132.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Phenylalanine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.