missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Proline, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.
Notes
Incompatible with strong oxidizing agents.
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼209°C (decomposition) |
Beilstein | 80809 |
Merck Index | 14,7780 |
Solubility Information | Soluble in water and absolute alcohol. Insoluble in ether,butanol and isopropanol. |
Formula Weight | 115.13 |
Percent Purity | 99% |
Quantity | 100 g |
Chemical Name or Material | DL-Proline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'