missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Proline, 99%

Catalog Number p-187559
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
609-36-9
C5H9NO2
115.132
MFCD00005250
ONIBWKKTOPOVIA-UHFFFAOYSA-N
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
614
CHEBI:26271
pyrrolidine-2-carboxylic acid
C1CC(NC1)C(=O)O
This item is not returnable. View return policy
609-36-9
C5H9NO2
115.132
MFCD00005250
ONIBWKKTOPOVIA-UHFFFAOYSA-N
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
614
CHEBI:26271
pyrrolidine-2-carboxylic acid
C1CC(NC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 208.0°C to 210.0°C
Color White
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Beilstein 22, 4
Fieser 09,393
Merck Index 15, 7895
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol, sparingly soluble in chloroform, benzene and, acetone, insoluble in ether
Residue after Ignition 0.1% max.
Formula Weight 115.13
Percent Purity 99%
Water 0.5% max. (K.F.)
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material DL-Proline, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'