missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Serine methyl ester hydrochloride, 99%

Product Code. 10194590 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10194590

Brand: Thermo Scientific Chemicals 207450100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C4H9NO3·HCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H9NO3·HCl
MDL Number MFCD00012593

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Beilstein 04, 512
Quantity 10 g
Formula Weight 155.58
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.