missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
£25.93 - £247.23
Chemical Identifiers
CAS | 133-37-9 |
---|---|
Molecular Formula | C4H6O6 |
Molecular Weight (g/mol) | 150.09 |
MDL Number | MFCD00071626 |
InChI Key | FEWJPZIEWOKRBE-UHFFFAOYNA-N |
Synonym | dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid |
PubChem CID | 875 |
ChEBI | CHEBI:15674 |
IUPAC Name | 2,3-dihydroxybutanedioic acid |
SMILES | OC(C(O)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10676924
|
Thermo Scientific Acros
137871000 |
100 g | Plastic bottle |
£25.93
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10487330
|
Thermo Scientific Acros
137875000 |
500 g | Plastic bottle |
£74.77
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915061
|
Thermo Scientific Acros
137870025 |
2.5 kg | Plastic bottle |
£247.23
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
133-37-9 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
875 | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00071626 | |
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid | |
CHEBI:15674 | |
OC(C(O)C(O)=O)C(O)=O |
Specifications
133-37-9 | |
White | |
C4H6O6 | |
MFCD00071626 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, insoluble in chloroform | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
CHEBI:15674 | |
99.5% | |
Authentic | |
Crystalline Powder |
200.0°C to 206.0°C | |
99.50% | |
HO2CCH(OH)CH(OH)CO2H | |
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid | |
875 | |
150.09 | |
210°C | |
0.5% max. | |
DL-Tartaric acid |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- DL-Tartaric acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only