missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10191-41-0 |
---|---|
Molecular Formula | C29H50O2 |
Molecular Weight (g/mol) | 430.72 |
MDL Number | MFCD00072051 |
InChI Key | GVJHHUAWPYXKBD-IEOSBIPESA-N |
Synonym | vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol |
PubChem CID | 14985 |
ChEBI | CHEBI:18145 |
IUPAC Name | (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol |
SMILES | CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418906
|
Thermo Scientific Alfa Aesar
A17039.18 |
50 g |
N/A
|
N/A | |||||
11428906
|
Thermo Scientific Alfa Aesar
A17039.30 |
250 g |
N/A
|
N/A | |||||
11408906
|
Thermo Scientific Alfa Aesar
A17039.0B |
1000 g |
N/A
|
N/A | |||||
Description
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.
Solubility
Miscible with chloroform, vegetable oils, ether, acetone and alcohol. Immiscible with water.
Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
Chemical Identifiers
10191-41-0 | |
430.72 | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
14985 | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol |
C29H50O2 | |
MFCD00072051 | |
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol | |
CHEBI:18145 | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
Specifications
10191-41-0 | |
0.950 g/mL | |
240°C (464°F) | |
C29H50O2 | |
MFCD00072051 | |
Air and light sensitive | |
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol | |
14985 | |
430.72 | |
DL-α-Tocopherol |
2°C to 4°C | |
200°C to 220°C (0.1 mm) | |
>97% | |
1.505 | |
94012 | |
14,10021 | |
Miscible with chloroform,vegetable oils, ether,acetone and alcohol. Immiscible with water. | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 | |
430.72 | |
CHEBI:18145 | |
≥97% |
Safety and Handling
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
missing translation for 'einecsNumber' : 233-466-0
missing translation for 'rtecsNumber' : GA8746000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DL-alpha-Tocopherol
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-tri methyltridecyl)-2H-1-benzopyran-6-ol
RUO – Research Use Only