missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Tryptophan, 99%

Product Code. 11332296
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11332296

Brand: Thermo Scientific Chemicals L05936.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Tryptophan, CAS # 54-12-6, is an α-amino acid that is commonly used in the synthesis of proteins. It is an essential amino acid precursor for the formation of serotonin, melatonin, and vitamin B3. | CAS: 54-12-6 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• DL-Tryptophan is an α-amino acid, and an essential amino acid precursor of serotonin, melatonin, and vitamin B3

Application

• DL-Tryptophan shows antioxidant activity• It has a role as a Daphnia magna metabolite

• It has been explored in in vitro studies to evaluate the antimicrobial and immunoregulatory functions on indoleamine 2,3-dioxygenase

• It has been used as a medium supplement in Psilocybe atrobrunnea and Psilocybe serbica for the synthesis of compounds

• It can be used as a medium supplement for culturing yeast

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-12-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064339
InChI Key QIVBCDIJIAJPQS-UHFFFAOYSA-N
Synonym dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
PubChem CID 1148
ChEBI CHEBI:27897
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∽280°C (decomposition)
Beilstein 86196
Solubility Information Soluble I water. 10g/L (20°C),Hcl and alkalies. Sparingly soluble in alcohol.
Quantity 25 g
Formula Weight 204.23
Percent Purity 99%
Chemical Name or Material DL-Tryptophan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.